衣康酸酐 ≥95.0% (by titrimetric analysis)
供应商: TCI
|
Warning
|
Synonyms:
2-亚甲基丁二酸酐
I0203-25G
I0203-500G
TCIAI0203-25GEA
0
CNY
TCIAI0203-25G
TCIAI0203-500G
衣康酸酐 ≥95.0% (by titrimetric analysis)
衣康酸酐
分子式:
C₅H₄O₃ 沸点: 115°C 溶点: 64…65 °C |
MDL编号:
MFCD00005530 CAS号: 2170-03-8 EINECS: 218-518-2 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (Morpholine Method) | min. 95.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...