2,2-二乙氧基丙烷 ≥95.0% (通过GC)

供应商: TCI
Danger

D0459-10ML D0459-250ML
TCIAD0459-10MLEA 0 CNY
TCIAD0459-10ML TCIAD0459-250ML
2,2-二乙氧基丙烷 ≥95.0% (通过GC)
2,2-二乙氧基丙烷
分子式: C₇H₁₆O₂
沸点: 113°C
密度: 0,826 g/cm³ (18 °C)
闪点: 7°C
存储条件: Refrigerator
MDL编号: MFCD00009224
CAS号: 126-84-1
EINECS: 204-808-6
UN号: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9498d7631abd687f783186232282804d6f03b5af&vid=9498d7631abd687f783186232282804d6f03b5af&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR