丙酮肟 ≥98.0% (by GC)
供应商: TCI
Synonyms:
Propan-2-one oxime, Acetoxime, Acetone oxime, 2-Propanone oxime
A0065-25G
A0065-500G
TCIAA0065-25GEA
0
CNY
TCIAA0065-25G
TCIAA0065-500G
丙酮肟 ≥98.0% (by GC)
丙酮肟
分子式:
C₃H₇NO 分子量: 73.09 g/mol 沸点: 133°C 溶点: 61 |
MDL编号:
MFCD00002118 CAS号: 127-06-0 EINECS: 204-820-1 Merck Index: 13,00077 |
|
产品技术规格
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 60.0 to 63.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...