3-二乙氨基-1-丙醇 ≥95.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 3-(Diethylamino)propan-1-ol, 3-Diethylamino-1-propanol

D0472-25ML D0472-500ML
TCIAD0472-25MLEA 0 CNY
TCIAD0472-25ML TCIAD0472-500ML
3-二乙氨基-1-丙醇 ≥95.0% (by GC, titration analysis)
3-二乙氨基-1-丙醇
分子式: C₇H₁₇NO
沸点: 189°C
密度: 0,8845 g/cm³ (20 °C)
闪点: 65°C
MDL编号: MFCD00002947
CAS号: 622-93-5
EINECS: 210-759-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f4fab5003e41fc3f970c204105870963e5007a9&vid=2f4fab5003e41fc3f970c204105870963e5007a9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR