三氟乙酸钠 ≥98.0% (通过滴定分析)

供应商: TCI
Danger

T1336-25G T1336-500G
TCIAT1336-25GEA 0 CNY
TCIAT1336-25G TCIAT1336-500G
三氟乙酸钠 ≥98.0% (通过滴定分析)
三氟乙酸钠
分子式: C₂F₃NaO₂
溶点: 207°C
MDL编号: MFCD00013217
CAS号: 2923-18-4
EINECS: 220-879-6
UN号: 2811
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c511646fbb8195c60c78a2a9c8f2ac6f6225f4b&vid=5c511646fbb8195c60c78a2a9c8f2ac6f6225f4b&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Water very faint turbidity
Water max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR