氨基甲酸甲酯 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
尿基烷
C1012-25G
C1012-500G
TCIAC1012-25GEA
0
CNY
TCIAC1012-25G
TCIAC1012-500G
氨基甲酸甲酯 ≥98.0% (by GC)
氨基甲酸甲酯
分子式:
C₂H₅NO₂ 沸点: 177 °C (1013 hPa) 溶点: 53…54 °C 密度: 1,1361 g/cm³ (56 °C) |
MDL编号:
MFCD00007964 CAS号: 598-55-0 EINECS: 209-939-2 Merck Index: 14,06036 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 53.0 to 58.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...