氨基甲酸丁酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Carbamic acid butyl ester

C0613-25G C0613-500G
TCIAC0613-25GEA 0 CNY
TCIAC0613-25G TCIAC0613-500G
氨基甲酸丁酯 ≥98.0% (by GC)
氨基甲酸丁酯
分子式: C₅H₁₁NO₂
沸点: 204 °C (1013 hPa)
溶点: 50…52 °C
MDL编号: MFCD00007967
CAS号: 592-35-8
EINECS: 209-751-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43afcb6cb6b6dac2b8e3b89ee77fb54ee927e057&vid=43afcb6cb6b6dac2b8e3b89ee77fb54ee927e057&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 57.0 deg-C
Solubility in Methanol within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR