2-丁氧基乙酸乙酯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 乙二醇丁醚乙酸酯, 乙酸-2-丁氧基乙酯

B0700-25ML B0700-500ML
TCIAB0700-25MLEA 0 CNY
TCIAB0700-25ML TCIAB0700-500ML
2-丁氧基乙酸乙酯 ≥98.0% (by GC)
2-丁氧基乙酸乙酯
分子式: C₈H₁₆O₃
沸点: 192°C
溶点: -64°C
密度: 0,9416 g/cm³ (20 °C)
闪点: 71°C
MDL编号: MFCD00009457
CAS号: 112-07-2
EINECS: 203-933-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e98a864dc2033be588e6f4790376c4b4a8816af&vid=0e98a864dc2033be588e6f4790376c4b4a8816af&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless clear liquid
Purity (GC) min. 98.0 %
Water max. 0.20 %
Acid Value max. 0.2


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR