棕榈酸甲酯 ≥97.0% (by GC)

供应商: TCI

Synonyms: 软脂酸甲酯, 十六酸甲酯

P0006-25G P0006-250G
TCIAP0006-25GEA 0 CNY
TCIAP0006-25G TCIAP0006-250G
棕榈酸甲酯 ≥97.0% (by GC)
棕榈酸甲酯
分子式: C₁₇H₃₄O₂
分子量: 270.46 g/mol
沸点: 417
溶点: 29.9
MDL编号: MFCD00008994
CAS号: 112-39-0
EINECS: 203-966-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=38f70d63a93c1fb0816597b9de059bc2e020250a&vid=38f70d63a93c1fb0816597b9de059bc2e020250a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to lump
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR