庚酸乙酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: 水芹酸乙酯

H0031-25ML H0031-500ML
TCIAH0031-25MLEA 0 CNY
TCIAH0031-25ML TCIAH0031-500ML
庚酸乙酯 ≥98.0% (by GC)
庚酸乙酯
分子式: C₉H₁₈O₂
分子量: 158.24 g/mol
沸点: 188°C
溶点: -66
闪点: 71°C
MDL编号: MFCD00009538
CAS号: 106-30-9
EINECS: 203-382-9
Merck Index: 14,03835
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02153c1499d70f35126367e28c25f2276557c8f3&vid=02153c1499d70f35126367e28c25f2276557c8f3&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR