2,3-丁二酮一肟 ≥98.0% (by GC)
供应商: TCI
Synonyms:
联乙酰一肟, 二乙酰一肟, BDM
B0683-25G
B0683-500G
TCIAB0683-25GEA
0
CNY
TCIAB0683-25G
TCIAB0683-500G
2,3-丁二酮一肟 ≥98.0% (by GC)
2,3-丁二酮一肟
分子式:
C₄H₇NO₂ 分子量: 101.11 g/mol 沸点: 186°C 溶点: 74 存储条件: Ambient |
MDL编号:
MFCD00002116 CAS号: 57-71-6 EINECS: 200-348-5 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 77.0 deg-C |
Solubility in hot Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...