3-溴-4-甲基噻吩 ≥93.0% (通过GC)

供应商: TCI

B1541-25G B1541-5G
TCIAB1541-25GEA 0 CNY
TCIAB1541-25G TCIAB1541-5G
3-溴-4-甲基噻吩 ≥93.0% (通过GC)
3-溴-4-甲基噻吩
分子式: C₅H₅BrS
沸点: 180°C
闪点: 77°C
MDL编号: MFCD00130084
CAS号: 30318-99-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7cea2bfcd740b94b6674e62ed6edb8cb04a10f32&vid=7cea2bfcd740b94b6674e62ed6edb8cb04a10f32&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 93.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR