2-乙酰基-1-甲基吡咯 ≥98.0% (by GC)
供应商: TCI
Synonyms:
1-(1-Methyl-1H-pyrrol-2-yl)ethanone, 2-Acetyl-1-methylpyrrole
A1014-25ML
A1014-5ML
TCIAA1014-25MLEA
0
CNY
TCIAA1014-25ML
TCIAA1014-5ML
2-乙酰基-1-甲基吡咯 ≥98.0% (by GC)
2-乙酰基-1-甲基吡咯
分子式:
C₇H₉NO 分子量: 123.15 g/mol 沸点: 202°C 闪点: 82°C 存储条件: Refrigerator |
MDL编号:
MFCD00003089 CAS号: 932-16-1 EINECS: 213-247-6 |
|
产品技术规格
Appearance | Colorless to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...