2-(2-吡啶基)乙胺 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 2-(2-氨乙基)吡啶

A1999-25G A1999-5G
TCIAA1999-25GEA 0 CNY
TCIAA1999-25G TCIAA1999-5G
2-(2-吡啶基)乙胺 ≥98.0% (by GC, titration analysis)
2-(2-吡啶基)乙胺
分子式: C₇H₁₀N₂
分子量: 122.17 g/mol
沸点: 93°C
存储条件: Ambient
MDL编号: MFCD00006367
CAS号: 2706-56-1
EINECS: 220-295-1
UN号: 2735
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ade54a800758f457330185444639b385bd023167&vid=ade54a800758f457330185444639b385bd023167&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR