尼克酰胺 ≥99.0% (by HPLC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
吡啶-3-甲酰胺, 维生素 B3, 烟酰胺, 烟碱酰胺, 维生素 PP
N0078-25G
N0078-500G
TCIAN0078-25GEA
0
CNY
TCIAN0078-25G
TCIAN0078-500G
尼克酰胺 ≥99.0% (by HPLC, titration analysis)
尼克酰胺
分子式:
C₆H₆N₂O 分子量: 122.13 g/mol 沸点: 334 闪点: 182 存储条件: Ambient |
MDL编号:
MFCD00006395 CAS号: 98-92-0 EINECS: 202-713-4 Merck Index: 14,06523 |
|
产品技术规格
Appearance | White powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 129.0 to 132.0 deg-C |
Solubility in Water | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...