烟酸乙酯 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
尼古丁酸乙酯
N0085-25ML
N0085-500ML
TCIAN0085-25MLEA
0
CNY
TCIAN0085-25ML
TCIAN0085-500ML
烟酸乙酯 ≥99.0% (by GC)
烟酸乙酯
分子式:
C₈H₉NO₂ 分子量: 151.16 g/mol 沸点: 224°C 闪点: 93°C 存储条件: Ambient |
MDL编号:
MFCD00006389 CAS号: 614-18-6 EINECS: 210-370-7 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...