三氯化钌

供应商: TCI
Danger

Synonyms: Ruthenium trichloride

R0074-1G R0074-5G
TCIAR0074-1GEA 0 CNY
TCIAR0074-1G TCIAR0074-5G
三氯化钌
三氯化钌
分子式: Cl₃Ru
溶点: 500 °C
密度: 3,11 g/cm³ (20 °C)
MDL编号: MFCD00011208
CAS号: 10049-08-8
EINECS: 233-167-5
UN号: 3260
ADR: 8,III
Merck Index: 14,08302
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85051b4636d3041d6e05c0cd9014d637a015474a&vid=85051b4636d3041d6e05c0cd9014d637a015474a&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Black powder to crystal
Content (Ruthenium) min. 40.0 %
Drying loss 1.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR