十甲基环五硅氧烷 ≥99.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Cyclopentasiloxane, 2,2,4,4,6,6,8,8,10,10-Decamethyl-1,3,5,7,9,2,4,6,8,10-pentaoxapentasilecane, Dow Corning 245® fluid, Volasil 245, Silicone fluid DC 245
D1890-25ML
D1890-500ML
TCIAD1890-25MLEA
0
CNY
TCIAD1890-25ML
TCIAD1890-500ML
十甲基环五硅氧烷 ≥99.0% (by GC)
十甲基环五硅氧烷
分子式:
C₁₀H₃₀O₅Si₅ 分子量: 370.77 g/mol 沸点: 210°C 溶点: –38°C 闪点: 76°C (closed cup) 存储条件: Ambient |
MDL编号:
MFCD00046966 CAS号: 541-02-6 EINECS: 208-764-9 Merck Index: 14,02848 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...