2-环己烯-1-醇 ≥95.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Cyclohex-2-ene-1-ol, Cyclohex-2-enol
C0816-25ML
C0816-5ML
TCIAC0816-25MLEA
0
CNY
TCIAC0816-25ML
TCIAC0816-5ML
2-环己烯-1-醇 ≥95.0% (by GC)
2-环己烯-1-醇
分子式:
C₆H₁₀O 分子量: 98.14 g/mol 沸点: 166°C 闪点: 58°C |
MDL编号:
MFCD00001570 CAS号: 822-67-3 EINECS: 212-501-3 UN号: 1987 ADR: 3 III |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...