2-环己基乙醇 ≥98.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(2-羟乙基)环己烷
C0582-25ML
C0582-500ML
TCIAC0582-25MLEA
0
CNY
TCIAC0582-25ML
TCIAC0582-500ML
2-环己基乙醇 ≥98.0% (by GC)
2-环己基乙醇
分子式:
C₈H₁₆O 分子量: 128.21 g/mol 沸点: 206°C 溶点: -20 闪点: 86°C 存储条件: Ambient |
MDL编号:
MFCD00001525 CAS号: 4442-79-9 EINECS: 224-672-1 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...