2′,4′,6′-三甲基苯乙酮 ≥97.0% (通过GC)

供应商: TCI

T1117-25ML T1117-5ML
TCIAT1117-25MLEA 0 CNY
TCIAT1117-25ML TCIAT1117-5ML
2′,4′,6′-三甲基苯乙酮 ≥97.0% (通过GC)
2′,4′,6′-三甲基苯乙酮
分子式: C₁₁H₁₄O
沸点: 236°C
密度: 0,975 g/cm³ (20 °C)
MDL编号: MFCD00008735
CAS号: 1667-01-2
EINECS: 216-783-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebbf8cf43ca50523ae7348c30ea646f54130d9f1&vid=ebbf8cf43ca50523ae7348c30ea646f54130d9f1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR