2,4,6-三甲基苯甲酰氯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
2,4,6-Trimethylbenzene-1-carbonyl chloride, 2,4,6-Trimethylbenzoyl Chloride
T2470-25G
T2470-500G
TCIAT2470-25GEA
0
CNY
TCIAT2470-25G
TCIAT2470-500G
2,4,6-三甲基苯甲酰氯 ≥98.0% (by GC, titration analysis)
2,4,6-三甲基苯甲酰氯
分子式:
C₁₀H₁₁ClO 沸点: 143°C 密度: 1,0967 g/cm³ (25 °C) 存储条件: Refrigerator |
MDL编号:
MFCD00013650 CAS号: 938-18-1 EINECS: 213-339-6 UN号: 3265 ADR: 8,II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...