2′,4′-二甲基苯乙酮 ≥95.0% (通过GC)

供应商: TCI

D1879-25ML D1879-5ML
TCIAD1879-25MLEA 0 CNY
TCIAD1879-25ML TCIAD1879-5ML
2′,4′-二甲基苯乙酮 ≥95.0% (通过GC)
2′,4′-二甲基苯乙酮
分子式: C₁₀H₁₂O
沸点: 122°C
密度: 1,0121 g/cm³ (15 °C)
闪点: 100°C
MDL编号: MFCD00003571
CAS号: 89-74-7
EINECS: 201-935-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b5c0032cb990be607734f817e083e9da464d33b9&vid=b5c0032cb990be607734f817e083e9da464d33b9&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR