1,2-苯二甲醇 ≥98.0% (by GC)

供应商: TCI

Synonyms: 1,2-双(羟基甲基)苯, 邻苯二甲醇, 邻二甲苯-α,α′-二醇

X0057-25G X0057-5G
TCIAX0057-25GEA 0 CNY
TCIAX0057-25G TCIAX0057-5G
1,2-苯二甲醇 ≥98.0% (by GC)
1,2-苯二甲醇
分子式: C₈H₁₀O₂
分子量: 138.17 g/mol
沸点: 145°C
MDL编号: MFCD00004626
CAS号: 612-14-6
EINECS: 210-293-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=288dd2399522bff6b381e51f165073a97642dfdf&vid=288dd2399522bff6b381e51f165073a97642dfdf&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 62.0 to 66.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR