4-叔丁基苯甲醇 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
(4-(tert-Butyl)phenyl)methanol, (4-tert-Butylphenyl)methanol
B1761-25G
B1761-5G
TCIAB1761-25GEA
0
CNY
TCIAB1761-25G
TCIAB1761-5G
4-叔丁基苯甲醇 ≥97.0% (by GC)
4-叔丁基苯甲醇
分子式:
C₁₁H₁₆O 沸点: 238°C 密度: 1,05 g/cm³ (20 °C) 闪点: 87°C 存储条件: Ambient |
MDL编号:
MFCD00004662 CAS号: 877-65-6 EINECS: 212-894-1 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...