3,5-双(三氟甲基)苯甲酰氯 ≥98.0% (通过滴定分析)

供应商: TCI
Danger

B2000-25G B2000-5G
TCIAB2000-25GEA 0 CNY
TCIAB2000-25G TCIAB2000-5G
3,5-双(三氟甲基)苯甲酰氯 ≥98.0% (通过滴定分析)
3,5-双(三氟甲基)苯甲酰氯
分子式: C₉H₃ClF₆O
沸点: 81°C
密度: 1,53 g/cm³ (19 °C)
闪点: 72°C
MDL编号: MFCD00000387
CAS号: 785-56-8
EINECS: 212-322-0
UN号: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc4915fab89f6c436790ef066fd12dd94b7046ae&vid=cc4915fab89f6c436790ef066fd12dd94b7046ae&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR