对甲苯磺酸钠 ≥90.0% (by titrimetric analysis)

供应商: TCI

Synonyms: 4-甲苯磺酸

T0271-500G T0271-25G
TCIAT0271-500GEA 0 CNY
TCIAT0271-500G TCIAT0271-25G
对甲苯磺酸钠 ≥90.0% (by titrimetric analysis)
对甲苯磺酸钠
分子式: C₇H₇NaO₃S
分子量: 194.19 g/mol
MDL编号: MFCD00064388
CAS号: 657-84-1
EINECS: 211-522-5
Merck Index: 14,09533
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c945c92c8cffa1641a88ebec6008d489da7df3e&vid=9c945c92c8cffa1641a88ebec6008d489da7df3e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Ion exchange titration) min. 90.0 % (calcd.on dried substance)
Solubility in Water almost transparency
Drying loss max. 5.0 %
Chloride max. 0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR