对甲苯磺酸甲酯 ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: Methyl 4-methylbenzenesulphonate, p-Toluenesulphonic acid methyl ester, Methyl p-toluenesulphonate, Toluene-4-sulphonic acid methyl ester

T0269-25G T0269-500G
TCIAT0269-25GEA 0 CNY
TCIAT0269-25G TCIAT0269-500G
对甲苯磺酸甲酯 ≥98.0% (by GC)
对甲苯磺酸甲酯
分子式: C₈H₁₀O₃S
分子量: 186.23 g/mol
沸点: 292
闪点: 130
存储条件: Ambient
MDL编号: MFCD00008417
CAS号: 80-48-8
EINECS: 201-283-5
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe3da8b8e36ac12f4da6b585354954c58e480bf1&vid=fe3da8b8e36ac12f4da6b585354954c58e480bf1&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR