邻甲苯磺酰胺 ≥98.0% (by HPLC, total nitrogen)

供应商: TCI
Warning

Synonyms: 2-Methylbenzene-1-sulphonamide, 2-Methylbenzenesulphonamide, o-Toluenesulphonamide

T0280-500G T0280-25G
TCIAT0280-500GEA 0 CNY
TCIAT0280-500G TCIAT0280-25G
邻甲苯磺酰胺 ≥98.0% (by HPLC, total nitrogen)
邻甲苯磺酰胺
分子式: C₇H₉NO₂S
沸点: 330 °C (1013 hPa)
溶点: 155…156 °C
存储条件: Ambient
MDL编号: MFCD00007934
CAS号: 88-19-7
EINECS: 201-808-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d76beb6168b92d38ecac0e7978edb4d1f62f2c6&vid=8d76beb6168b92d38ecac0e7978edb4d1f62f2c6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 155.0 to 159.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR