2-羟基苄醇 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 邻羟基苄醇, 水杨醇

H0223-25G H0223-250G
TCIAH0223-25GEA 0 CNY
TCIAH0223-25G TCIAH0223-250G
2-羟基苄醇 ≥98.0% (by GC)
2-羟基苄醇
分子式: C₇H₈O₂
分子量: 124.14 g/mol
沸点: 268
溶点: 82
存储条件: Ambient
MDL编号: MFCD00004617
CAS号: 90-01-7
EINECS: 201-960-5
Merck Index: 14,08325
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73bdf1956d0177f4dc9495e096085762b0446c81&vid=73bdf1956d0177f4dc9495e096085762b0446c81&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light red powder to crystal
Purity (GC) min. 98.0 %
Melting point 84.0 to 87.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR