4-氨基苯甲醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
4-氨基苄醇
A1096-25G
A1096-5G
TCIAA1096-25GEA
0
CNY
TCIAA1096-25G
TCIAA1096-5G
4-氨基苯甲醇 ≥98.0% (by GC, titration analysis)
4-氨基苯甲醇
分子式:
C₇H₉NO 分子量: 123.15 g/mol 沸点: 171°C 溶点: 75 存储条件: Freezer |
MDL编号:
MFCD00014782 CAS号: 623-04-1 EINECS: 210-767-5 |
|
产品技术规格
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 61.0 to 65.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...