3-氨基苯甲醇 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
Synonyms:
3-氨基苄醇
A2259-25G
A2259-5G
TCIAA2259-25GEA
0
CNY
TCIAA2259-25G
TCIAA2259-5G
3-氨基苯甲醇 ≥98.0% (by GC, titration analysis)
3-氨基苯甲醇
分子式:
C₇H₉NO 分子量: 123.15 g/mol 沸点: 272 存储条件: Refrigerator |
MDL编号:
MFCD00007817 CAS号: 1877-77-6 EINECS: 217-514-8 |
|
产品技术规格
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...