2-氟-3-(三氟甲基)苯甲酸 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Warning
|
F0619-1G
F0619-5G
TCIAF0619-1GEA
0
CNY
TCIAF0619-1G
TCIAF0619-5G
2-氟-3-(三氟甲基)苯甲酸 ≥98.0% (by GC, titration analysis)
2-氟-3-(三氟甲基)苯甲酸
分子式:
C₈H₄F₄O₂ 分子量: 208.11 g/mol |
MDL编号:
MFCD00040980 CAS号: 115029-22-6 |
|
产品技术规格
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 125.0 to 129.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...