3′,4′-二氯苯丙酮 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 1-(3,4-Dichlorophenyl)-1-propanone, 1-(3,4-Dichlorophenyl)propan-1-one

D3365-25G D3365-5G
TCIAD3365-25GEA 0 CNY
TCIAD3365-25G TCIAD3365-5G
3′,4′-二氯苯丙酮 ≥98.0% (by GC)
3′,4′-二氯苯丙酮
分子式: C₉H₈Cl₂O
分子量: 203.07 g/mol
沸点: 247°C
溶点: 44
MDL编号: MFCD00000554
CAS号: 6582-42-9
EINECS: 229-511-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be4fc3a9f590dd8854ad99108e22b5597c5c4a37&vid=be4fc3a9f590dd8854ad99108e22b5597c5c4a37&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR