α-Bromo-2-chloro-4-fluorotoluene ≥98.0% (by GC)

供应商: TCI
Danger

Synonyms: 1-(Bromomethyl)-2-chloro-4-fluorobenzene, 2-Chloro-4-fluorobenzyl bromide

C2098-25G C2098-5G
TCIAC2098-25GEA 0 CNY
TCIAC2098-25G TCIAC2098-5G
α-Bromo-2-chloro-4-fluorotoluene ≥98.0% (by GC)
α-Bromo-2-chloro-4-fluorotoluene
分子式: C₇H₅BrClF
分子量: 223.47 g/mol
存储条件: Refrigerator
MDL编号: MFCD00236025
CAS号: 45767-66-6
UN号: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=506d84189b33a4574dc2651be652f51f7c0a4678&vid=506d84189b33a4574dc2651be652f51f7c0a4678&prid=web&clid=SAPDAM

现在订购

产品技术规格

Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR