对苯二异硫氰酸酯 ≥98.0% (by GC)
供应商: TCI
|
Danger
|
Synonyms:
双硫氰苯, 1,4-亚苯基二硫代异氰酸酯, PDITC
P1279-25G
P1279-5G
TCIAP1279-25GEA
0
CNY
TCIAP1279-25G
TCIAP1279-5G
对苯二异硫氰酸酯 ≥98.0% (by GC)
对苯二异硫氰酸酯
分子式:
C₈H₄N₂S₂ 沸点: 319 °C (1013 hPa) 溶点: 131 °C 存储条件: Refrigerator |
MDL编号:
MFCD00004811 CAS号: 4044-65-9 EINECS: 223-741-3 UN号: 2811 ADR: 6.1,III Merck Index: 14,01308 |
|
产品技术规格
Appearance | White to Green to Brown powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 129.0 to 133.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...