4-碘苯乙醚 ≥98.0% (by GC)

供应商: TCI

Synonyms: 对碘苯乙醚

I0525-25G I0525-5G
TCIAI0525-25GEA 0 CNY
TCIAI0525-25G TCIAI0525-5G
4-碘苯乙醚 ≥98.0% (by GC)
4-碘苯乙醚
分子式: C₈H₉IO
MDL编号: MFCD00061124
CAS号: 699-08-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba25b9f216449f0a9b6fa9fa2c4c272fe872e2b5&vid=ba25b9f216449f0a9b6fa9fa2c4c272fe872e2b5&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR