4-(三氟甲氧基)苯磺酰氯 ≥98.0% (by GC, titration analysis)
供应商: TCI
|
Danger
|
Synonyms:
4-(Trifluoromethoxy)benzene-1-sulphonyl chloride
T2146-25G
T2146-5G
TCIAT2146-25GEA
0
CNY
TCIAT2146-25G
TCIAT2146-5G
4-(三氟甲氧基)苯磺酰氯 ≥98.0% (by GC, titration analysis)
4-(三氟甲氧基)苯磺酰氯
分子式:
C₇H₄ClF₃O₃S 分子量: 260.62 g/mol 密度: 1.536 |
MDL编号:
MFCD00042408 CAS号: 94108-56-2 EINECS: 302-397-9 UN号: 3265 ADR: 8 II |
|
产品技术规格
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...