4-(三氟甲氧基)碘苯 ≥98.0% (by GC)

供应商: TCI
Warning

Synonyms: 4-碘-α,α,α-三氟甲氧基苯, 1-碘-4-三氟甲氧基苯

T2153-25G T2153-5G
TCIAT2153-25GEA 0 CNY
TCIAT2153-25G TCIAT2153-5G
4-(三氟甲氧基)碘苯 ≥98.0% (by GC)
4-(三氟甲氧基)碘苯
分子式: C₇H₄F₃IO
分子量: 288.01 g/mol
沸点: 97°C
闪点: 78°C
MDL编号: MFCD00042411
CAS号: 103962-05-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af71670fd6cecfb89a6059ebf1cd512b42f0b25e&vid=af71670fd6cecfb89a6059ebf1cd512b42f0b25e&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Yellow to Orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR