2-氟-6-羟基苯甲酸 ≥98.0% (by GC, titration analysis)

供应商: TCI
Warning

Synonyms: 6-氟水杨酸

F0553-1G F0553-5G
TCIAF0553-1GEA 0 CNY
TCIAF0553-1G TCIAF0553-5G
2-氟-6-羟基苯甲酸 ≥98.0% (by GC, titration analysis)
2-氟-6-羟基苯甲酸
分子式: C₇H₅FO₃
分子量: 156.11 g/mol
MDL编号: MFCD00052179
CAS号: 67531-86-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=89d1225489b0fe1d8bb17c1c4ff4dceaab694da6&vid=89d1225489b0fe1d8bb17c1c4ff4dceaab694da6&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 162.0 to 166.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR