2,6-Difluoro-4-hydroxybenzoic acid ≥98.0% (by GC)
供应商: TCI
|
Warning
|
D3985-1G
D3985-5G
TCIAD3985-1GEA
0
CNY
TCIAD3985-1G
TCIAD3985-5G
2,6-Difluoro-4-hydroxybenzoic acid ≥98.0% (by GC)
2,6-Difluoro-4-hydroxybenzoic acid
分子式:
C₇H₄F₂O₃ 分子量: 174.1 g/mol 溶点: 176°C |
MDL编号:
MFCD03094500 CAS号: 214917-68-7 |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...