对氟苯甲酸甲酯 ≥97.0% (by GC)
供应商: TCI
|
Warning
|
Synonyms:
Methyl 4-fluorobenzoate, 4-Fluorobenzoic acid emthyl ester
F0403-25G
F0403-500G
TCIAF0403-25GEA
0
CNY
TCIAF0403-25G
TCIAF0403-500G
对氟苯甲酸甲酯 ≥97.0% (by GC)
对氟苯甲酸甲酯
分子式:
C₈H₇FO₂ 分子量: 154.14 g/mol 沸点: 197°C 闪点: 77°C |
MDL编号:
MFCD00017959 CAS号: 403-33-8 EINECS: 206-956-7 |
|
产品技术规格
Appearance | Colorless to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...