3-硝基苯甲酸甲酯 ≥98.0% (by GC)

供应商: TCI

Synonyms: Methyl 3-Nitrobenzoate, 3-Nitrobenzoic acid methyl ester

N0157-25G N0157-500G
TCIAN0157-25GEA 0 CNY
TCIAN0157-25G TCIAN0157-500G
3-硝基苯甲酸甲酯 ≥98.0% (by GC)
3-硝基苯甲酸甲酯
分子式: C₈H₇NO₄
分子量: 181.15 g/mol
MDL编号: MFCD00007250
CAS号: 618-95-1
EINECS: 210-573-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e104c157e62945b50689cc9da35f6245a19a3a3f&vid=e104c157e62945b50689cc9da35f6245a19a3a3f&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR