3-氯苯磺酰氯 ≥98.0% (by GC, titration analysis)

供应商: TCI
Danger

Synonyms: 3-Chlorobenzene-1-sulphonyl chloride

C1791-1G C1791-5G
TCIAC1791-1GEA 0 CNY
TCIAC1791-1G TCIAC1791-5G
3-氯苯磺酰氯 ≥98.0% (by GC, titration analysis)
3-氯苯磺酰氯
分子式: C₆H₄Cl₂O₂S
分子量: 211.07 g/mol
沸点: 135°C
MDL编号: MFCD00051697
CAS号: 2888-06-4
EINECS: 220-753-0
UN号: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52cf897833c12d65dfbc9c055a30ec52d9b34700&vid=52cf897833c12d65dfbc9c055a30ec52d9b34700&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR