2,4,6-三氯苯磺酰氯 ≥98.0% (通过GC,滴定分析)
供应商: TCI
|
Danger
|
T2035-25G
T2035-5G
TCIAT2035-25GEA
0
CNY
TCIAT2035-25G
TCIAT2035-5G
2,4,6-三氯苯磺酰氯 ≥98.0% (通过GC,滴定分析)
2,4,6-三氯苯磺酰氯
分子式:
C₆H₂Cl₄O₂S 溶点: 49 °C |
MDL编号:
MFCD00052973 CAS号: 51527-73-2 UN号: 3261 ADR: 8,II |
|
产品技术规格
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Melting point | 48.0 to 51.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...