Ethyl-3-chlorobenzoate ≥94.0% (by GC)
供应商: TCI
Synonyms:
3-Chlorobenzoic acid ethyl ester, Ethyl 3-chlorobenzoate
C1737-25G
C1737-5G
TCIAC1737-25GEA
0
CNY
TCIAC1737-25G
TCIAC1737-5G
Ethyl-3-chlorobenzoate ≥94.0% (by GC)
Ethyl-3-chlorobenzoate
分子式:
C₉H₉ClO₂ 分子量: 184.62 g/mol |
MDL编号:
MFCD00013634 CAS号: 1128-76-3 EINECS: 214-441-3 |
|
产品技术规格
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...