4-溴苄肼 ≥97.0% (by titrimetric analysis)

供应商: TCI
Warning

Synonyms: 4-Bromobenzene-1-carbohydrazide, 4-Bromobenzoic acid hydrazide, 4-Bromobenzohydrazide, 4-Bromobenzoic hydrazide

B2067-25G B2067-5G
TCIAB2067-25GEA 0 CNY
TCIAB2067-25G TCIAB2067-5G
4-溴苄肼 ≥97.0% (by titrimetric analysis)
4-溴苄肼
分子式: C₇H₇BrN₂O
分子量: 215.05 g/mol
MDL编号: MFCD00007602
CAS号: 5933-32-4
EINECS: 227-681-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=521151fb8332aa282ef87459c64412f1c0be9015&vid=521151fb8332aa282ef87459c64412f1c0be9015&prid=web&clid=SAPDAM

现在订购

产品技术规格

Appearance White to Almost white powder to crystal
Purity (Bromination Method) min. 97.0 %
Melting point 166.0 to 169.0 deg-C
Solubility in hot Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

更多 About VWR