反式-2-己烯-1-醇 ≥95.0% (通过GC)
供应商: TCI
|
Warning
|
H0346-25G
H0346-250G
TCIAH0346-25GEA
0
CNY
TCIAH0346-25G
TCIAH0346-250G
反式-2-己烯-1-醇 ≥95.0% (通过GC)
反式-2-己烯-1-醇
分子式:
C₆H₁₂O 沸点: 162°C 溶点: 65…67 °C 密度: 0,8459 g/cm³ (20 °C) 闪点: 61°C 存储条件: Refrigerator |
MDL编号:
MFCD00002927 CAS号: 928-95-0 EINECS: 213-191-2 ADR: 3,III |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...