cis-4-Hepten-1-ol ≥95.0% (by GC)
供应商: TCI
H0833-25ML
H0833-5ML
TCIAH0833-25MLEA
0
CNY
TCIAH0833-25ML
TCIAH0833-5ML
cis-4-Hepten-1-ol ≥95.0% (by GC)
cis-4-Hepten-1-ol
分子式:
C₇H₁₄O 分子量: 114.19 g/mol 沸点: 85°C 闪点: 73°C |
MDL编号:
MFCD00015575 CAS号: 6191-71-5 EINECS: 228-238-2 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...