10-十一烯-1-醇 ≥98.0% (by GC)
供应商: TCI
Synonyms:
十一烯醇
U0047-25ML
U0047-500ML
TCIAU0047-25MLEA
0
CNY
TCIAU0047-25ML
TCIAU0047-500ML
10-十一烯-1-醇 ≥98.0% (by GC)
10-十一烯-1-醇
分子式:
C₁₁H₂₂O 分子量: 170.3 g/mol 沸点: 138°C 溶点: -1 闪点: 125°C |
MDL编号:
MFCD00004750 CAS号: 112-43-6 EINECS: 203-971-0 |
|
产品技术规格
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...